Learn More
Description
In vivo, ASP 7967 upregulates the serum hEPO concentration by 7.2-fold over the vehicle control (mice). Cell permeable and low-toxic, ASP 7967 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N2,N4-Bis(4-fluorophenyl)-N6-(2-pyrimidinylmethyl)-1,3,5-triazine-2,4,6-triamine hemifumarate |
| CAS | 1246958-42-8 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C20H16F2N8.½C4H4O4 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
