Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPA Antibody (OTI3G5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270216C
Description
ASPA Monoclonal specifically detects ASPA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ASPA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ASPA | |
Recombinant protein fragment corresponding to amino acids 77-313 of human ASPA (NP_000040) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3G5 | |
Western Blot, Immunohistochemistry | |
ACY-2, ACY2aminoacylase 2, Aminoacylase-2, ASPaminoacylase-2, aspartoacylase, aspartoacylase (aminoacylase 2, Canavan disease), EC 3.5.1.15 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
443.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction