Learn More
ASPA Mouse anti-Human, Clone: OTI3G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805303

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.Specifications
ASPA | |
Monoclonal | |
Unconjugated | |
ASPA | |
ACY2, ASP | |
Mouse | |
Affinity Chromatography | |
RUO | |
443 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P45381 | |
ASPA | |
Human recombit protein fragment corresponding to amino acids 77-313 of human ASPA produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.