Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

asparagine synthetase Mouse anti-Human, Clone: Z5808, MilliporeSigma™

Catalog No. MABS1300MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1300MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1300MI Supplier MilliporeSigma Supplier No. MABS1300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-asparagine synthetase, clone Z5808 Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting for the detection of human Asparagine Synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen asparagine synthetase
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone Z5808
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ASNS,TS11
Host Species Mouse
Immunogen N-asparagine synthetase-budded virus.
Purification Method Ammonium Sulfate
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001664
Test Specificity Clone Z5808 detects Asparagine synthetase in human creast cancer, human tonsil, and human uterus. It targets an epitope within 50 amino acids from the N-terminal end.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.