Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Asparagine Synthetase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556113 Supplier Invitrogen™ Supplier No. PA556113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHFEFEYQTK VDGEIILHLY DKGGIEQTIC MLDGVFAFVL LDTANKKVFL GRDTYGVRPL FKAMTEDGFL AVCSEAKGLV TLKHSATPFL KVEPFLPGHY EVLDLKPNGK VASVEMVKYH HCRDVP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asparagine Synthetase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Asns
Gene Accession No. P08243
Gene Alias ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein
Gene Symbols Asns
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ASNS (Product #RP-88728)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.