Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Asparagine synthetase Rabbit anti-Human, Mouse, Rat, Clone: 7M10Z7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Asparagine synthetase |
---|---|
Clone | 7M10Z7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Asparagine synthetase Monoclonal antibody specifically detects Asparagine synthetase in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Asparagine synthetase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
440 | |
IgG | |
Affinity purified |
7M10Z7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein | |
A synthetic peptide corresponding to a sequence within amino acids 450-550 of human Asparagine synthetase (P08243). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title