Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspartame, Powder, NF, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600799
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

A1378, 22839-47-0, C14H18N2O5

Spectrum™ Chemical Aspartame, Powder, NF is used in pharmaceutical products, often as a sugar replacement in chewable tablets and sugar-free liquids. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
IUPAC Name (3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 4.5%
Packaging Poly Pail
Specific Rotation +14.5 to +16.5°
Quantity 500 g
Percent Purity 98 to 102%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.