Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspartame, Powder, NF, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
22839-47-0
C14H18N2O5
294.31
IAOZJIPTCAWIRG-UHFFFAOYNA-N
3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid
COC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)CC(O)=O
22839-47-0
C14H18N2O5
294.31
IAOZJIPTCAWIRG-UHFFFAOYNA-N
3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid
COC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)CC(O)=O

A1378, 22839-47-0, C14H18N2O5

Spectrum™ Aspartame, Powder, NF is used in pharmaceutical products, often as a sugar replacement in chewable tablets and sugar-free liquids. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.045
Packaging Fiber Drum
Specific Rotation +14.5 to +16.5°
Percent Purity 98 to 102%
Quantity 12 kg