Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspartate Aminotransferase Antibody (3G11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259483F
This item is not returnable.
View return policy
Description
Aspartate Aminotransferase Monoclonal antibody specifically detects Aspartate Aminotransferase in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Aspartate Aminotransferase | |
Monoclonal | |
FITC | |
aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A | |
Recombinant human Aspartate Aminotransferase (1-413aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
2805 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, ELISA, Immunofluorescence | |
3G11 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction