Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspartate Aminotransferase Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093403
Description
Aspartate Aminotransferase Polyclonal specifically detects Aspartate Aminotransferase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Aspartate Aminotransferase | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
aspartate aminotransferase, cytoplasmic, EC 2.6.1.1, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), growth-inhibiting protein 18, Transaminase A | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 1 ug/ml, Immunohistochemistry 2 - 5 ug/ml, Immunohistochemistry-Paraffin 2 - 5 ug/ml, Peptide ELISA Detection limit 1:4000 | |
NP_002070.1 | |
GOT1 | |
Peptide with sequence C-RSYRYWDAEKR corresponding to internal region according to NP_002070.1. | |
0.1 mg | |
Lipid and Metabolism | |
2805 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction