Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspartate Transaminase Antibody, DyLight 405, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP178089V
Description
Aspartate Transaminase Polyclonal antibody specifically detects Aspartate Transaminase in Porcine samples. It is validated for Western Blot, ELISASpecifications
Aspartate Transaminase | |
Polyclonal | |
Western Blot, ELISA | |
aspartate aminotransferase, cytoplasmic, CASPAT | |
Aspartate Transaminase Antibody was produced by repeated immunizations with Aspartate Transaminase from pig heart. (Uniprot: P00503) | |
0.1 mL | |
Primary | |
Pig | |
IgG |
Western Blot, ELISA | |
DyLight 405 | |
50mM Sodium Borate | |
Sheep | |
Multi-step | |
RUO | |
396967 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction