Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspartate Transaminase Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991246
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1246 25 μL
50-199-1245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1246 Supplier Rockland Immunochemicals Supplier No. 200601144S
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Aspartate Transaminase is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep Serum as well as purified and partially purified Aspartate Transaminase (AST) [Pig Heart]. Cross reactivity against Aspartate Transaminase (AST) from other tissues and species may occur but have not been specifically determined.

GOT1 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.

Specifications

Antigen Aspartate Transaminase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene GOT1
Gene Accession No. P00503
Gene Alias AI789014; aspartate aminotransferase 1; Aspartate aminotransferase, cytoplasmic; aspartate transaminase 1; Aspat; AST1; ASTQTL1; cAspAT; cCAT; Cysteine aminotransferase, cytoplasmic; Cysteine transaminase, cytoplasmic; cytosolic aspartate aminotransferase; Gaspat; GIG18; Glutamate oxaloacetate transaminase 1; glutamate oxaloacetate transaminase 1, soluble; glutamic-oxaloacetic transaminase 1; glutamic-oxaloacetic transaminase 1, soluble; Got1; Got-1; growth-inhibiting protein 18; testis secretory sperm-binding protein Li 196a; Transaminase A
Gene Symbols GOT1
Host Species Sheep
Immunogen Anti-Aspartate Transaminase antibody was produced by repeated immunizations with Aspartate Transaminase from pig heart.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396967
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.