Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aspartoacylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582793 Supplier Invitrogen™ Supplier No. PA582793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AIIHPNLQDQ DWKPLHPGDP MFLTLDGKTI PLGGDCTVYP VFVNEAAYYE KKEAFAKTTK LTLNAK Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.

ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspartoacylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASPA
Gene Accession No. P45381
Gene Alias ACY2; ACY-2; aminoacylase 2; aminoacylase-2; ASP; ASPA; Aspartoacylase; aspartoacylase (aminoacylase) 2; nur7
Gene Symbols ASPA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASPA. Recombinant protein control fragment (Product #RP-93187).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.