Learn More
Aspartyl-tRNA Synthetase/DARS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Aspartyl-tRNA Synthetase/DARS |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8709
![]() |
Bethyl Laboratories, Inc
A304799AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.Specifications
Aspartyl-tRNA Synthetase/DARS | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
aspRS, aspartate tRNA ligase 1, cytoplasmic, aspartate--tRNA ligase, cytoplasmic, aspartyl-tRNA synthetase, cytoplasmic, cell proliferation-inducing gene 40 protein, cell proliferation-inducing protein 40 | |
Between 25 and 75 | |
Primary | |
4° C | |
DARS |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P14868, Q922B2 | |
1615, 226414 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.