Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPDH Antibody (OTI4G8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272508AF750
Description
ASPDH Monoclonal specifically detects ASPDH in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
ASPDH | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ASPDH | |
Full length recombinant protein of human ASPDH (NP_001019827) produced in E.coli. | |
0.1 mL | |
554235.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4G8 | |
Immunohistochemistry | |
aspartate dehydrogenase domain containing, Aspartate dehydrogenase domain-containing protein, EC 1.4.1.21, putative L-aspartate dehydrogenase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction