Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPDH Monoclonal Antibody (OTI4G8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527041

Catalog No. PIMA527041


Only null left
Add to Cart

Description

Description

ASPDH Monoclonal Antibody for IHC (P)

Specifically catalyzes the NAD or NADP-dependent dehydrogenation of L-aspartate to iminoaspartate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ASPDH
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
A6ND91
ASPDH
Full length human recombinant protein of ASPDH produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
OTI4G8
Unconjugated
ASPDH
0610012D14Rik; aspartate dehydrogenase domain containing; aspartate dehydrogenase domain-containing protein; Aspdh; Putative L-aspartate dehydrogenase; RGD1310111
Mouse
Affinity Chromatography
RUO
554235
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.