Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570197 Supplier Invitrogen™ Supplier No. PA570197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 92%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rat: 93%.

Specifically catalyzes the NAD or NADP-dependent dehydrogenation of L-aspartate to iminoaspartate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPDH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASPDH
Gene Accession No. A6ND91
Gene Alias 0610012D14Rik; aspartate dehydrogenase domain containing; aspartate dehydrogenase domain-containing protein; Aspdh; Putative L-aspartate dehydrogenase; RGD1310111
Gene Symbols ASPDH
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human ASPDH.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 554235
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.