Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aspergillus Recombinant Rabbit Monoclonal Antibody (5145)
GREENER_CHOICE

Catalog No. PIMA533312
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533312 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533312 Supplier Invitrogen™ Supplier No. MA533312
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Reactive with 9 Aspergillus species including 6 strains of A. fumigatus.

Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspergillus
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5145
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.5
Host Species Rabbit
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.