Learn More
Invitrogen™ Aspergillus Polyclonal Antibody, Biotin

Description
The PA1-73166 antibody reacts with Aspergillus species. PA1-73166 has been successfully used in immunofluorescence applications. The PA1-73166 antibody was raised against soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus. Recommended short term storage of less than 6 months at 2-8ºC. For extended storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Aspergillus |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus. |
| Purification Method | IgG fraction |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.