Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aspergillus Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA173165
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173165 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173165 Supplier Invitrogen™ Supplier No. PA173165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73165 antibody reacts with Aspergillus species. PA1-73165 has been successfully used in ELISA applications. The PA1-73165 antibody was raised against soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus. Short term storage, up to 6 months, is liquid at 4°C. For longer term storage, aliquot and freeze.

Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspergillus
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1-2 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Host Species Rabbit
Immunogen Soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.