Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543688 Supplier Invitrogen™ Supplier No. PA543688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SEVLQGKLGI YDADGDGDFD VDDAKVLLEG PSGVAKRKTK AKVKELTKEE Sequence homology: Dog: 93%; Horse: 83%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 86%.

ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins. This gene is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPH
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Asph
Gene Accession No. Q12797
Gene Alias 2310005F16Rik; 3110001L23Rik; A beta H-J-J; AAH; AI848629; ASP beta-hydroxylase; Aspartate beta-hydroxylase; aspartate-beta-hydroxylase; aspartyl (asparaginyl) beta hydroxylase; aspartyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase-like; aspartyl/asparaginyl-beta-hydroxylase; ASPH; AW261690; AW561901; BAH; C79816; calsequestrin-binding protein; cardiac junctin; CASQ2BP1; cI-37; FDLAB; HAAH; humbug; JCTN; jumbug; junctate; junctin; junctional sarcoplasmic reticulum protein; peptide-aspartate beta-dioxygenase; Unknown (protein for MGC:137539)
Gene Symbols Asph
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ASPH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 444
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.