Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspirin, USP, 99.5-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Aspirin, USP is a salicylate drug that is used as an analgesic, an antipyretic, and an anti-inflammatory. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Ignition Residue | 0.05% |
Loss on Drying | 0.5% |
Packaging | Poly Bottle |
Quantity | 500 g |
UN Number | UN2811 |
Percent Purity | 99.5 to 100.5% |
Grade | USP |
Physical Form | Crystalline Powder or Lumps |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction