Learn More
ASPL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302351AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.Specifications
ASPL | |
Polyclonal | |
Unconjugated | |
ASPSCR1 | |
UBX domain protein 9; UBX domain-containing protein 9; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; alveolar soft part sarcoma locus; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; tether containing UBX domain for GLUT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79058 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9BZE9 | |
ASPSCR1 | |
Between 175 and 225 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.