Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Asporin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114883 Supplier Invitrogen™ Supplier No. PA5114883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Asporin.

ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asporin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Aspn
Gene Accession No. Q99MQ4, Q9BXN1
Gene Alias 4631401G09Rik; AA986886; ASPN; Asporin; asporin (LRR class 1); asporin proteoglycan; FLJ20129; OS3; periodontal ligament associated protein 1; periodontal ligament-associated protein 1; PLAP1; PLAP-1; SLRR1C; small leucine-rich protein 1C; UNQ215/PRO241
Gene Symbols Aspn
Host Species Rabbit
Immunogen A synthesized peptide derived from human ASPN(Accession Q9BXN1), corresponding to amino acid residues L311-P361.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306805, 54829, 66695
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.