Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASPP1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB246357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB246357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB246357 Supplier Novus Biologicals Supplier No. NB11040684AF700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASPP1 Polyclonal antibody specifically detects ASPP1 in Human samples. It is validated for Western Blot

Specifications

Antigen ASPP1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias apoptosis-stimulating protein of p53, 1, ASPP1Protein phosphatase 1 regulatory subunit 13B, KIAA0771apoptosis-stimulating of p53 protein 1, p53BP2-like, p85, protein phosphatase 1, regulatory (inhibitor) subunit 13B
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human Apoptosis-Stimulated Protein of p53, 1 using the numbering given in entry NP_056131.1 (GeneID 23368).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23368
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.