Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPP1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040684DL594
Description
ASPP1 Polyclonal antibody specifically detects ASPP1 in Human samples. It is validated for Western BlotSpecifications
ASPP1 | |
Polyclonal | |
Western Blot | |
apoptosis-stimulating protein of p53, 1, ASPP1Protein phosphatase 1 regulatory subunit 13B, KIAA0771apoptosis-stimulating of p53 protein 1, p53BP2-like, p85, protein phosphatase 1, regulatory (inhibitor) subunit 13B | |
The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human Apoptosis-Stimulated Protein of p53, 1 using the numbering given in entry NP_056131.1 (GeneID 23368). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23368 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction