Learn More
ASPP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300918A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the ASPP family of p53 interacting proteins. The protein contains four ankyrin repeats and an SH3 domain involved in protein-protein interactions. ASPP proteins are required for the induction of apoptosis by p53-family proteins. They promote DNA binding and transactivation of p53-family proteins on the promoters of proapoptotic genes. Expression of this gene is regulated by the E2F transcription factor.Specifications
| ASPP1 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R13B | |
| Protein phosphatase 1, regulatory subunit 13B, ASPP1, KIAA0771, p53BP2-like, p85 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23368 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96KQ4 | |
| PPP1R13B | |
| Between 475 and 525 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.