Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASPP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982412
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2412 Supplier Bioss Supplier No. BS1282R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the ASPP family of p53 interacting proteins. The protein contains four ankyrin repeats and an SH3 domain involved in protein-protein interactions. ASPP proteins are required for the induction of apoptosis by p53-family proteins. They promote DNA binding and transactivation of p53-family proteins on the promoters of proapoptotic genes. Expression of this gene is regulated by the E2F transcription factor.

Specifications

Antigen ASPP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PPP1R13B
Gene Accession No. Q96KQ4
Gene Alias AI449786; Apoptosis-stimulating of p53 protein 1; apoptosis-stimulating protein of p53, 1; Aspp1; AW545810; KIAA0771; p53BP2-like; p85; Ppp1r13b; Protein phosphatase 1 regulatory subunit 13B; protein phosphatase 1, regulatory (inhibitor) subunit 13B; protein phosphatase 1, regulatory subunit 13B; Tp53bp2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein, 2; tumor protein p53-binding protein, 2
Gene Symbols PPP1R13B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASPP1 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23368
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.