Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Asprosin Antibody (Birdy-1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311677G
Description
Asprosin Monoclonal specifically detects Asprosin in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
Asprosin | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
>95%, by SDS-PAGE | |
RUO | |
2200 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA | |
Birdy-1 | |
Western Blot, ELISA | |
FBN1, fibrillin 1 | |
Recombinant human asprosin. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction