Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asprosin Antibody (Birdy-1), Janelia Fluor 669, R&D Systems™
SDP

Catalog No. NB523873 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB523873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB523873 Supplier R&D Systems Supplier No. NBP311677JF669

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Asprosin Monoclonal antibody specifically detects Asprosin in Human,Mouse samples. It is validated for ELISA,Western Blot

Specifications

Antigen Asprosin
Applications ELISA, Western Blot
Classification Monoclonal
Clone Birdy-1
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias FBN1, fibrillin 1
Host Species Mouse
Immunogen Recombinant human asprosin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2200
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.