Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Asprosin Recombinant Rabbit Monoclonal Antibody (RM463)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544582
Description
Asprosin is an hormone derived from the Fibrillin-1 which corresponds to residues from 2734 to 2871 of the Fibrillin protein.
Asprosin is a protein hormone produced by mammals in tissues that stimulates the liver to release glucose into the blood stream. Asprosin is encoded by the gene FBN1 as part of the protein profibrillin and is released from the C-terminus of the latter by specific proteolysis.
Specifications
Asprosin | |
Recombinant Monoclonal | |
50 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P35555 | |
Rabbit | |
Protein A | |
RUO | |
2200 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
RM463 | |
Unconjugated | |
FBN1 | |
FBN1 | |
Recombinant full-length human Asprosin protein which correspond to the full length protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction