Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AspRS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555323
Description
Immunogen sequence: EVTLCGWIQY RRQNTFLVLR DFDGLVQVII PQDESAASVK KILCEAPVES VVQVSGTVIS RPAGQENPKM PTGEIEIKVK TAELLNACKK LPFEIKNFVK K Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. It is a mitochondrial enzyme that specifically aminoacylates aspartyl-tRNA. Mutations in this gene are associated with leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation (LBSL).
Specifications
AspRS | |
Polyclonal | |
Unconjugated | |
DARS2 | |
5830468K18Rik; aspartate tRNA ligase 2, mitochondrial; aspartate--tRNA ligase, mitochondrial; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase 2 (mitochondrial); aspartyl-tRNA synthetase 2, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartate--tRNA ligase, mitochondrial; aspRS; Dars2; LBSL; MT-ASPRS; RGD1308286; si:dkey-21n10.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PI48 | |
DARS2 | |
Recombinant protein corresponding to Human AspRS. Recombinant protein control fragment (Product #RP-94128). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction