Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AspRS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598518
Description
AspRS Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. It is a mitochondrial enzyme that specifically aminoacylates aspartyl-tRNA. Mutations in this gene are associated with leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation (LBSL).
Specifications
AspRS | |
Polyclonal | |
Unconjugated | |
DARS2 | |
5830468K18Rik; aspartate tRNA ligase 2, mitochondrial; aspartate--tRNA ligase, mitochondrial; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase 2 (mitochondrial); aspartyl-tRNA synthetase 2, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartyl-tRNA synthetase, mitochondrial; aspartate--tRNA ligase, mitochondrial; aspRS; Dars2; LBSL; MT-ASPRS; RGD1308286; si:dkey-21n10.1 | |
Rabbit | |
Protein G | |
RUO | |
55157 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6PI48 | |
DARS2 | |
Recombinant Human Aspartate--tRNA ligase, mitochondrial protein (489-628AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction