Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPRV1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP233981
Description
ASPRV1 Polyclonal specifically detects ASPRV1 in Human, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ASPRV1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q53RT3 | |
ASPRV1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GKLKLKAQFLVANASAEEAIIGTDVLQDHNAILDFEHRTCTLKGKKFRLLPVGGSLEDEFDLELIEEDPSSEEGRQELSH | |
0.1 mL | |
Proteases & Other Enzymes | |
151516 | |
Human, Canine | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
aspartic peptidase, retroviral-like 1, EC 3.4.23.-, FLJ25084, retroviral-like aspartic protease 1, SASPaseMUNO, SASPTAPS, Skin aspartic protease, Skin-specific retroviral-like aspartic protease, Taps, TPA-inducible aspartic proteinase-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction