Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASPRV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557070
Description
Immunogen sequence: GKLKLKAQFL VANASAEEAI IGTDVLQDHN AILDFEHRTC TLKGKKFRLL PVGGSLEDEF DLELIEEDPS SEEGRQELSH Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 30%.
Filaggrin is a structural protein that is crucial for in the development and maintenance of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.
Specifications
ASPRV1 | |
Polyclonal | |
Unconjugated | |
ASPRV1 | |
2300003P22Rik; AA986851; aspartic peptidase retroviral like 1; aspartic peptidase, retroviral-like 1; asprv1; LOW QUALITY PROTEIN: retroviral-like aspartic protease 1; MUNO; Retroviral-like aspartic protease 1; RGD1560859; SASP; SASPase; skin aspartic protease; skin-specific retroviral-like aspartic protease; TAPS; TPA-inducible aspartic proteinase-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53RT3 | |
ASPRV1 | |
Recombinant protein corresponding to Human ASPRV1. Recombinant protein control fragment (Product #RP-96209). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction