Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPRV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557071 Supplier Invitrogen™ Supplier No. PA557071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHLPKEIVFA NSMGKGYYLK GKIGKVPVRF LVDSGAQVSV VHPNLWEEVT DGDLDTLQPF ENVVKVANGA EMKILGVWDT AVSL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 25%.

Filaggrin is a structural protein that is crucial for in the development and maintenance of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPRV1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASPRV1
Gene Accession No. Q53RT3
Gene Alias 2300003P22Rik; AA986851; aspartic peptidase retroviral like 1; aspartic peptidase, retroviral-like 1; asprv1; LOW QUALITY PROTEIN: retroviral-like aspartic protease 1; MUNO; Retroviral-like aspartic protease 1; RGD1560859; SASP; SASPase; skin aspartic protease; skin-specific retroviral-like aspartic protease; TAPS; TPA-inducible aspartic proteinase-like protein
Gene Symbols ASPRV1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASPRV1. Recombinant protein control fragment (Product #RP-96210).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151516
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.