Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASPRV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569911
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
Filaggrin is a structural protein that is crucial for in the development and maintenance of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.
Specifications
ASPRV1 | |
Polyclonal | |
Unconjugated | |
ASPRV1 | |
2300003P22Rik; AA986851; aspartic peptidase retroviral like 1; aspartic peptidase, retroviral-like 1; asprv1; LOW QUALITY PROTEIN: retroviral-like aspartic protease 1; MUNO; Retroviral-like aspartic protease 1; RGD1560859; SASP; SASPase; skin aspartic protease; skin-specific retroviral-like aspartic protease; TAPS; TPA-inducible aspartic proteinase-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67855 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q09PK2 | |
ASPRV1 | |
synthetic peptide directed towards the middle region of mouse ASPRV1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction