Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPRV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113990 Supplier Invitrogen™ Supplier No. PA5113990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGEALGVYNR LSPQDQGDYG TVKEALLKAF GVPGAAPSHL PKEIVFANSM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Guinea Pig: 92%; Horse: 100%; Human: 100%; Rat: 93%.

Filaggrin is a structural protein that is crucial for in the development and maintenance of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPRV1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASPRV1
Gene Accession No. Q53RT3
Gene Alias 2300003P22Rik; AA986851; aspartic peptidase retroviral like 1; aspartic peptidase, retroviral-like 1; asprv1; LOW QUALITY PROTEIN: retroviral-like aspartic protease 1; MUNO; Retroviral-like aspartic protease 1; RGD1560859; SASP; SASPase; skin aspartic protease; skin-specific retroviral-like aspartic protease; TAPS; TPA-inducible aspartic proteinase-like protein
Gene Symbols ASPRV1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SASP.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151516
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.