Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPRV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 130521AP150UL

Description
Filaggrin is a structural protein that is crucial for in the development and maintece of the skin barrier. This gene encodes a retroviral-like protease involved in profilaggrin-to-filaggrin processing. Expression is found primarily in the epidermis and inner root sheath of hair follicles.Specifications
ASPRV1 | |
Polyclonal | |
Unconjugated | |
ASPRV1 | |
ASPRV1, MUNO, SASP, SASPase, Skin aspartic protease, Taps | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151516, 500243, 67855 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q09PK2, Q53RT3 | |
Asprv1 | |
ASPRV1 Fusion Protein Ag3719 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction