Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASPZODRAFT_1728931 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144587 Supplier Invitrogen™ Supplier No. PA5144587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASPZODRAFT_1728931 Polyclonal Antibody for Western Blot, ELISA

ASPZODRAFT_1728931 is an identifier for a predicted gene or protein sequence that has been identified in the genome of Aspergillus zonatus, a species of filamentous fungus. The specific function and characteristics of ASPZODRAFT_1728931 are not yet known, and further research would be required to elucidate its role in cellular processes and its potential implications for fungal physiology and ecology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPZODRAFT_1728931
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias ASPZODRAFT_1728931
Host Species Rabbit
Immunogen Recombinant Penicilliopsis zonata CBS 506.65 ASPZODRAFT_1728931 protein (173-274aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.