Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AsRed2 Mouse anti-Tag, HRP, Clone: OTI2E2, liquid, TrueMAB™
SDP

Catalog No. 501671170
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1170 Supplier OriGene Supplier No. TA183022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AsRed2 fluorescent protein.

Specifications

Antigen AsRed2
Applications Western Blot
Classification Monoclonal
Clone OTI2E2
Concentration 0.5 mg/mL
Conjugate HRP
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias AsRed2
Host Species Mouse
Immunogen AsRed2 recombit protein expressed in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.