Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASRGL1 Antibody (CRASH/1290), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308327F
Description
ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for ELISASpecifications
ASRGL1 | |
Monoclonal | |
FITC | |
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase | |
Recombinant human ASRGL1 fragment (Uniprot: Q7L266) | |
100 μg | |
Neuroscience | |
80150 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
CRASH/1290 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction