Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASRGL1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100155
Description
ASRGL1 Polyclonal specifically detects ASRGL1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
ASRGL1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Both reported variants represent identical protein (NP_001077395.1 and NP_079356.3). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_001077395.1 | |
ASRGL1 | |
Peptide with sequence EKHEKGAQKTDCQ corresponding to internal region according to NP_001077395.1, NP_079356.3. | |
0.1 mg | |
Neuroscience | |
80150 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction