Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASS1 Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809138

Description
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.Specifications
ASS1 | |
Monoclonal | |
Unconjugated | |
ASS1 | |
ASS, CTLN1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
445 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI4G9 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P00966 | |
ASS1 | |
Synthetic peptide corresponding to residues near C-terminus of human ASS1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction