Learn More
ASS1 Mouse anti-Human, Clone: OTI8A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809139

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.Specifications
ASS1 | |
Monoclonal | |
Unconjugated | |
ASS1 | |
ASS, CTLN1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
445 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P00966 | |
ASS1 | |
Synthetic peptide corresponding to residues near C-terminus of human ASS1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.