Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572474
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572474 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572474 Supplier Invitrogen™ Supplier No. PA572474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASS1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ASS1
Gene Accession No. P00966, P09034, P16460
Gene Alias AA408052; arginino succinate synthetase; argininosuccinate synthase; argininosuccinate synthase 1; argininosuccinate synthetase; argininosuccinate synthetase 1; arginosuccinate synthetase 1; ASS; Ass1; Ass-1; ASSA; citrulline-aspartate ligase; citrulline--aspartate ligase; citrullinemia; CN; CTLN1; fold; mutant arginino succinate synthetase; RP11-618A20.2
Gene Symbols ASS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 192-221 amino acids from the Central region of human ASS.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11898, 25698, 445
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.