Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ 18 in. Chemical Resistance Nitrile Gloves, Sandpatch Grip

Catalog No. 19152501 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-152-501 Large
19-152-500 Medium
19-152-502 X-Large
19-152-503 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-152-501 Supplier PIP™ Supplier No. 50N2272G/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unsupported, unlined 22 mil nitrile gloves are resistant to physical and chemical hazards such as cut, puncture, solvents and oil based chemicals.

  • Made from component materials that comply with Federal Regulations for food contact
  • Unlined, chlorinated
  • Straight cuff
  • Color: green
  • Sandpatch grip
  • Overall Length: 18 in. (46 cm)
  • Ideal for:
    • Chemical processing
    • Oil refining and petrochemicals
    • Food processing
    • Aerospace and automotive degreasing
    • Automotive assembly and painting

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE, EN 374-3
Chlorinated Yes
Applications Industrial
Color Green
Finger Thickness (English) 22 mil
Finger Thickness (Metric) 0.56 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 22 mil
Palm Thickness (Metric) 0.56 mm
Product Type Chemical-resistant Gloves
Size Large
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
EN388 Standard EN 388:4102
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES AJK
For Use With (Application) Chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
Glove Cuff Style Straight
Grip Design Sandpatch
Lining Unlined
Model 50-N2272G/L
Series Assurance™ 50-N2272G
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.