Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Green 15 mil Flock-Lined Chemical Resistant Nitrile Gloves

Catalog No. 19152508 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-152-508 Large
19-152-507 Medium
19-152-509 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-152-508 Supplier PIP™ Supplier No. 50N160G/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unsupported lined gloves provide protection from chemical and physical hazards. Commonly used in light chemical applications, printing and meat processing.

  • Resistant to cut, puncture, snag and abrasion
  • Resistant to many chemical hazards, including solvents and oil-based chemicals
  • Lightweight construction and hand specific design provide comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge nitriles
  • Raised diamond grip pattern
  • Chlorinated
  • Straight cuff, Length: 13 in. (33 cm)
  • Thickness: 15 mil
  • Green

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE, EN 374-3
Chlorinated No
Applications Industrial
Color Green
Finger Thickness (English) 15 mil
Finger Thickness (Metric) 0.39 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 15 mil
Palm Thickness (Metric) 0.38 mm
Product Type Chemical-resistant Gloves
Size Large
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:4101
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES AJK
For Use With (Application) Chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
Glove Cuff Style Straight
Grip Design Raised Diamond
Liner Material Cotton Flock
Lining Lined
Model 50-N160G/L
Series Assurance™ 50-N160B
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.