Learn More
Medicom™ AssureMask™ Balance™ Earloop Face Masks
Catalog No. 17000271 Shop All Medicom Products
Click to view available options
Color:
Blue
Pink
White
Yellow
ASTM Level:
ASTM Level 2
ASTM Level 3
This item is not returnable. View return policy
This item is not returnable. View return policy
Face masks provide healthcare professionals and other frontline workers with the ideal balance between protection and comfort without gapping or irritation.
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Adjustable nosepiece ensures a comfortable, customized fit
  • Ultrasonically sealed, soft, stretchy earloops are free of natural rubber latex, preventing pulling and irritation
  • Made from high-quality materials that provide reliable protection without compromising breathability
  • Assorted colors available
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 2
Color Yellow
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes
Model 205517
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.