Learn More
Medicom™ AssureMask Precision™ Tie-On Face Masks
Tie-on face masks provide ASTM Level 1, 2 or 3 protection. Made from high quality materials that enhance breathability, these masks have soft, ample-length ties, an adjustable nose piece and a soft inner lining to ensure a comfortable, customized fit.
Supplier:  Medicom™ 206818
 View more versions of this product
Catalog No. 17-000-564
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Non-woven spunbond outer layer, provides maximum protection against fluid penetration
  • Ample length ties are soft, strong, ultrasonically sealed and attached to the outside of the mask to prevent pulling and irritation
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High quality media ensures optimal filtration and breathability
  • SureView™ Technology Fog-free nose strip ensures an unobstructed view of the operative field
Specifications
Specifications
Medical/Procedural Face Mask
Spunbond Polypropylene
Non-sterile
Single-use
Yes
50 Masks/Box, 6 Boxes/Cs.
Ties
Standard
Pleated
≥95% (PFE), ≥95% (BFE)
206818
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.